Assay Details
Target Gene Details
Entrez Gene ID: | 57587 |
Gene Name: | cilia and flagella associated protein 97 |
Gene Aliases: |
KIAA1430, hmw |
Location: |
Chr.4:185159662-185210251 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFAP97 | NM_001292033.1 | NP_001278962.1 | ||
| NM_020827.2 | NP_065878.1 | |||
| XM_017008483.1 | XP_016863972.1 | |||
| XM_017008484.1 | XP_016863973.1 | |||
| XM_017008485.1 | XP_016863974.1 | |||
| XM_017008486.1 | XP_016863975.1 | |||
| AB037851.1 | BAA92668.1 | |||
| AK091993.1 | ||||
| AL133615.1 | CAB63743.1 | |||
| BC030535.1 | AAH30535.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv461859 | Chr.4:184907523 - 185314394 on Build GRCh38 | Loss |
|

Genomic Map