Assay Details
Target Gene Details
Entrez Gene ID: | 5158 |
Gene Name: | phosphodiesterase 6B |
Gene Aliases: |
CSNB3, CSNBAD2, PDEB, RP40, rd1 |
Location: |
Chr.4:587593-670892 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE6B | NM_000283.3 | NP_000274.2 | ||
| NM_001145291.1 | NP_001138763.1 | |||
| XM_011513473.2 | XP_011511775.1 | |||
| XM_011513474.2 | XP_011511776.1 | |||
| XM_011513475.2 | XP_011511777.1 | |||
| XM_011513476.2 | XP_011511778.1 | |||
| BC000249.1 | AAH00249.1 | |||
| BT009794.1 | AAP88796.1 | |||
| S41458.1 | AAB22690.1 | |||
| X66142.1 | CAA46932.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759217 | Chr.4:609419 - 937566 on Build GRCh38 | Loss |
|
| nsv822425 | Chr.4:546316 - 1420011 on Build GRCh38 | Loss |
|
| nsv593205 | Chr.4:608002 - 629840 on Build GRCh38 | Loss |
|
| nsv950325 | Chr.4:620112 - 796912 on Build GRCh38 | Deletion |
|
| nsv508986 | Chr.4:527899 - 756994 on Build GRCh38 | Insertion |
|
| esv993737 | Chr.4:624323 - 635048 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115501488] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map