Assay Details
Target Gene Details
Entrez Gene ID: | 323 |
Gene Name: | amyloid beta precursor protein binding family B member 2 |
Gene Aliases: |
FE65L, FE65L1 |
Location: |
Chr.4:40810027-41216714 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv10486 | Chr.4:40747595 - 40935088 on Build GRCh38 | Gain |
|
| dgv1477e212 | Chr.4:40820049 - 40825701 on Build GRCh38 | Gain |
|
| nsv829912 | Chr.4:40720092 - 40840600 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map