Assay Details
Target Gene Details
Entrez Gene ID: | 8310 |
Gene Name: | acyl-CoA oxidase 3, pristanoyl |
Gene Aliases: |
- |
Location: |
Chr.4:8366263-8440725 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACOX3 | NM_001101667.1 | NP_001095137.1 | ||
| NM_003501.2 | NP_003492.2 | |||
| XM_005248011.4 | XP_005248068.1 | |||
| XM_005248013.3 | XP_005248070.1 | |||
| XM_011513565.2 | XP_011511867.1 | |||
| XM_011513566.1 | XP_011511868.1 | |||
| AK225080.1 | ||||
| AK313242.1 | ||||
| BC017053.1 | AAH17053.1 | |||
| Y11411.1 | CAA72214.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv593637 | Chr.4:8352102 - 8388919 on Build GRCh38 | Loss |
|
| nsv593638 | Chr.4:8371217 - 8456667 on Build GRCh38 | Loss |
|
| nsv1002794 | Chr.4:8108356 - 8489207 on Build GRCh38 | Gain |
|
| esv1005399 | Chr.4:8385554 - 8396256 on Build GRCh38 | Deletion |
|
| nsv1009011 | Chr.4:8223942 - 8423104 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map