Assay Details
Target Gene Details
Entrez Gene ID: | 101410542 |
Gene Name: | UCHL1 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.4:41220074-41256727 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UCHL1-AS1 | NR_102709.1 | |||
| DB169394.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3600330 | Chr.4:41225366 - 41230057 on Build GRCh38 | Loss |
|
| esv3563761 | Chr.4:41225545 - 41230233 on Build GRCh38 | Deletion |
|
| nsv829914 | Chr.4:41072681 - 41242403 on Build GRCh38 | Loss |
|

Genomic Map