Assay Details
Target Gene Details
Entrez Gene ID: | 9429 |
Gene Name: | ATP binding cassette subfamily G member 2 (Junior blood group) |
Gene Aliases: |
ABC15, ABCP, BCRP, BCRP1, BMDP, CD338, CDw338, EST157481, GOUT1, MRX, MXR, MXR-1, MXR1, UAQTL1 |
Location: |
Chr.4:88090264-88231417 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCG2 | NM_001257386.1 | NP_001244315.1 | ||
| NM_004827.2 | NP_004818.2 | |||
| XM_005263354.3 | XP_005263411.1 | |||
| XM_005263355.3 | XP_005263412.1 | |||
| XM_005263356.3 | XP_005263413.1 | |||
| XM_011532420.2 | XP_011530722.1 | |||
| XM_017008852.1 | XP_016864341.1 | |||
| XM_017008853.1 | XP_016864342.1 | |||
| AB051855.1 | BAB46933.1 | |||
| AB056867.1 | BAB39212.1 | |||
| AF093771.1 | ||||
| AF093772.1 | ||||
| AF098951.2 | AAC97367.1 | |||
| AF103796.1 | AAD09188.1 | |||
| AF463519.1 | AAO14617.1 | |||
| AK002040.1 | BAA92050.1 | |||
| AK290000.1 | ||||
| AY017168.1 | AAG52982.1 | |||
| AY288307.1 | AAP31310.1 | |||
| AY289766.1 | AAP44087.1 | |||
| AY333755.1 | AAQ92941.1 | |||
| AY333756.1 | AAQ92942.1 | |||
| BC021281.2 | AAH21281.1 | |||
| BC092408.1 | AAH92408.1 | |||
| U66681.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4413 | Chr.4:88083754 - 88129196 on Build GRCh38 | Deletion |
|
| nsv441897 | Chr.4:88102936 - 88160287 on Build GRCh38 | Gain |
|
| esv2665331 | Chr.4:87937174 - 88153583 on Build GRCh38 | Deletion |
|
| esv2759266 | Chr.4:87997399 - 88291967 on Build GRCh38 | Gain |
|
| nsv10531 | Chr.4:88099214 - 88163241 on Build GRCh38 | Gain |
|
| esv3601296 | Chr.4:88100712 - 88148969 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map