Assay Details
Target Gene Details
Entrez Gene ID: | 116449 |
Gene Name: | cytokine dependent hematopoietic cell linker |
Gene Aliases: |
MIST |
Location: |
Chr.4:10489899-10734853 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLNK | NM_052964.2 | NP_443196.2 | ||
| XM_011513775.2 | XP_011512077.1 | |||
| XM_017007684.1 | XP_016863173.1 | |||
| XM_017007685.1 | XP_016863174.1 | |||
| AB032369.1 | BAA96241.1 | |||
| AB110420.1 | BAC76765.1 | |||
| AK093001.1 | ||||
| BC137553.1 | ||||
| BC137556.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4234 | Chr.4:10511797 - 10545696 on Build GRCh38 | Insertion |
|
| nsv517971 | Chr.4:10397444 - 10891629 on Build GRCh38 | Gain |
|
| nsv593727 | Chr.4:10395703 - 10693367 on Build GRCh38 | Gain |
|
| esv2759223 | Chr.4:10342075 - 10544176 on Build GRCh38 | Loss |
|
| esv3599624 | Chr.4:10476161 - 10518729 on Build GRCh38 | Loss |
|
| nsv1014335 | Chr.4:9435268 - 10687720 on Build GRCh38 | Gain |
|
| esv3893876 | Chr.4:10345427 - 10660766 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116306449] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map