Assay Details
Target Gene Details
Entrez Gene ID: | 150159 |
Gene Name: | solute carrier family 9 member B1 |
Gene Aliases: |
NHA1, NHEDC1 |
Location: |
Chr.4:102885048-103019739 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC9B1 | NM_001100874.2 | NP_001094344.1 | ||
| NM_139173.3 | NP_631912.2 | |||
| NR_047513.1 | ||||
| NR_047515.1 | ||||
| XM_006714093.3 | XP_006714156.1 | |||
| XM_011531622.1 | XP_011529924.1 | |||
| AF447585.1 | AAM22868.1 | |||
| AK313592.1 | ||||
| AY461581.1 | ||||
| BC110794.1 | AAI10795.1 | |||
| BC132712.1 | ||||
| BC136966.1 | ||||
| DQ003059.1 | AAY21808.1 |
Target Gene Details
Entrez Gene ID: | 133308 |
Gene Name: | solute carrier family 9 member B2 |
Gene Aliases: |
NHA2, NHE10, NHEDC2 |
Location: |
Chr.4:103018033-103077371 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC9B2 | XM_006714085.2 | 13 | 3948 | XP_006714148.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4445 | Chr.4:103003411 - 103034963 on Build GRCh38 | Insertion |
|
| nsv594978 | Chr.4:102886563 - 103032232 on Build GRCh38 | Loss |
|
| esv2759270 | Chr.4:102859548 - 103136164 on Build GRCh38 | Gain |
|
| nsv830021 | Chr.4:103013297 - 103179431 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79723528] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map