Assay Details
Target Gene Details
Entrez Gene ID: | 57587 |
Gene Name: | cilia and flagella associated protein 97 |
Gene Aliases: |
KIAA1430, hmw |
Location: |
Chr.4:185159662-185210251 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFAP97 | NM_020827.2 | 5 | 4518 | NP_065878.1 |
| XM_017008483.1 | 5 | 4776 | XP_016863972.1 | |
| XM_017008484.1 | 5 | 4410 | XP_016863973.1 | |
| XM_017008485.1 | 7 | 4643 | XP_016863974.1 | |
| XM_017008486.1 | 5 | 4410 | XP_016863975.1 | |
| AK024878.1 | 1 | 402 | ||
| AV709118.2 | 1 | 246 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv461859 | Chr.4:184907523 - 185314394 on Build GRCh38 | Loss |
|

Genomic Map