Assay Details
Target Gene Details
Entrez Gene ID: | 4214 |
Gene Name: | mitogen-activated protein kinase kinase kinase 1 |
Gene Aliases: |
MAPKKK1, MEKK, MEKK 1, MEKK1, SRXY6 |
Location: |
Chr.5:56815073-56896152 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP3K1 | NM_005921.1 | NP_005912.1 | ||
| AF042838.1 | AAC97073.1 | |||
| AK307828.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv23998 | Chr.5:56814909 - 56816627 on Build GRCh38 | Loss |
|
| nsv598205 | Chr.5:56815393 - 56831971 on Build GRCh38 | Gain |
|
| dgv9789n54 | Chr.5:56815030 - 56818699 on Build GRCh38 | Gain |
|
| dgv9790n54 | Chr.5:56815165 - 56816579 on Build GRCh38 | Gain |
|
| dgv9792n54 | Chr.5:56815342 - 56816579 on Build GRCh38 | Loss |
|
| nsv598208 | Chr.5:56815495 - 56818023 on Build GRCh38 | Gain |
|
| nsv830308 | Chr.5:56601676 - 56823403 on Build GRCh38 | Gain |
|
| dgv9791n54 | Chr.5:56815268 - 56816476 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map