Assay Details
Target Gene Details
Entrez Gene ID: | 8878 |
Gene Name: | sequestosome 1 |
Gene Aliases: |
A170, FTDALS3, OSIL, PDB3, ZIP3, p60, p62, p62B |
Location: |
Chr.5:179806388-179838078 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SQSTM1 | NM_001142298.1 | NP_001135770.1 | ||
| NM_001142299.1 | NP_001135771.1 | |||
| XM_011534683.2 | 3 | 1125 | XP_011532985.1 | |
| AK093109.1 | 3 | 1715 | ||
| AK096241.1 | ||||
| AK226167.1 | ||||
| BC001874.1 | AAH01874.1 | |||
| BI549805.1 | ||||
| DA541885.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469579 | Chr.5:179757830 - 179933191 on Build GRCh38 | Loss |
|
| nsv600668 | Chr.5:179800865 - 179815581 on Build GRCh38 | Loss |
|
| esv3570596 | Chr.5:179804973 - 179813403 on Build GRCh38 | Loss |
|
| esv988291 | Chr.5:179806076 - 179813737 on Build GRCh38 | Deletion |
|
| nsv823361 | Chr.5:179786323 - 179824243 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs555215408,rs73351626] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map