Assay Details
Target Gene Details
Entrez Gene ID: | 54514 |
Gene Name: | DEAD-box helicase 4 |
Gene Aliases: |
VASA |
Location: |
Chr.5:55737994-55817146 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX4 | NM_001142549.1 | NP_001136021.1 | ||
| NM_001166533.1 | NP_001160005.1 | |||
| NM_024415.2 | NP_077726.1 | |||
| XM_011543495.2 | XP_011541797.1 | |||
| XM_011543497.2 | XP_011541799.1 | |||
| AF262962.1 | AAF72705.1 | |||
| AK292417.1 | ||||
| AK308300.1 | ||||
| AY004154.1 | AAF86585.1 | |||
| BC047455.1 | AAH47455.1 | |||
| BC088362.1 | AAH88362.1 | |||
| BI559479.1 | ||||
| DB457419.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830306 | Chr.5:55673052 - 55846654 on Build GRCh38 | Gain |
|
| nsv830305 | Chr.5:55543776 - 55743734 on Build GRCh38 | Loss |
|
| esv2759343 | Chr.5:55685067 - 55763164 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map