Assay Details
Target Gene Details
Entrez Gene ID: | 100652758 |
Gene Name: | uncharacterized LOC100652758 |
Gene Aliases: |
- |
Location: |
Chr.5:151769793-151772176 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100652758 | NR_132345.1 | |||
| NR_132346.1 | ||||
| AB593120.1 | ||||
| BI461699.1 | ||||
| CN293193.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs116550045] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map