Assay Details
Target Gene Details
Entrez Gene ID: | 6940 |
Gene Name: | zinc finger protein 354A |
Gene Aliases: |
EZNF, HEL104, HKL1, KID-1, KID1, TCF17 |
Location: |
Chr.5:178711521-178730702 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF354A | NM_001324339.1 | NP_001311268.1 | ||
| NM_005649.2 | NP_005640.2 | |||
| XM_011534645.2 | XP_011532947.1 | |||
| XM_017009791.1 | XP_016865280.1 | |||
| AF116030.1 | AAD05335.1 | |||
| AK290711.1 | ||||
| BC047105.1 | AAH47105.1 | |||
| D89928.1 | BAA25182.1 | |||
| EU794664.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3607724 | Chr.5:178707314 - 178726984 on Build GRCh38 | Gain |
|
| esv3890769 | Chr.5:178698920 - 178955866 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map