Assay Details
Target Gene Details
Entrez Gene ID: | 102157402 |
Gene Name: | adenylate kinase 6 |
Gene Aliases: |
AD-004, CGI-137, CINAP, CIP, hCINAP |
Location: |
Chr.5:69351726-69370013 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AK6 | NM_001015891.1 | NP_001015891.1 | ||
| NM_016283.4 | NP_057367.1 | |||
| AF110777.1 | AAF14860.1 | |||
| AF151895.1 | AAD34132.1 | |||
| AJ878880.1 | CAI48030.1 | |||
| AJ878881.1 | CAI48031.1 | |||
| BC007426.2 | AAH07426.1 | |||
| BX117839.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5694n100 | Chr.5:69081291 - 69374274 on Build GRCh38 | Gain |
|
| nsv821643 | Chr.5:69272838 - 71076658 on Build GRCh38 | Loss |
|
| nsv428117 | Chr.5:69347454 - 71500057 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map