Assay Details
Target Gene Details
Entrez Gene ID: | 285600 |
Gene Name: | KIAA0825 |
Gene Aliases: |
C5orf36 |
Location: |
Chr.5:94150851-94618604 on Build GRCh38 |
Assay Gene Location: | Within Intron 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIAA0825 | NM_001145678.1 | NP_001139150.1 | ||
| XM_006714588.2 | XP_006714651.1 | |||
| AK130941.1 | BAC85469.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2138679 | Chr.5:94172966 - 94173410 on Build GRCh38 | Deletion |
|
| nsv327963 | Chr.5:94172700 - 94182250 on Build GRCh38 | Deletion |
|

Genomic Map