Assay Details
Target Gene Details
Entrez Gene ID: | 4214 |
Gene Name: | mitogen-activated protein kinase kinase kinase 1 |
Gene Aliases: |
MAPKKK1, MEKK, MEKK 1, MEKK1, SRXY6 |
Location: |
Chr.5:56815073-56896152 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP3K1 | NM_005921.1 | NP_005912.1 | ||
| XM_017009484.1 | 1 | 2525 | XP_016864973.1 | |
| XM_017009485.1 | XP_016864974.1 | |||
| AF042838.1 | AAC97073.1 | |||
| AK307828.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv598205 | Chr.5:56815393 - 56831971 on Build GRCh38 | Gain |
|
| nsv830308 | Chr.5:56601676 - 56823403 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72022199,rs73759210] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map