Assay Details
Target Gene Details
Entrez Gene ID: | 285588 |
Gene Name: | EF-hand calcium binding domain 9 |
Gene Aliases: |
- |
Location: |
Chr.5:172188061-172203454 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFCAB9 | NM_001171183.1 | NP_001164654.1 | ||
| XM_017009350.1 | XP_016864839.1 | |||
| AI024830.1 | ||||
| CN445899.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759392 | Chr.5:172113419 - 172308268 on Build GRCh38 | Gain+Loss |
|
| nsv1015251 | Chr.5:172166251 - 172261560 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map