Assay Details
Target Gene Details
Entrez Gene ID: | 51149 |
Gene Name: | chromosome 5 open reading frame 45 |
Gene Aliases: |
- |
Location: |
Chr.5:179837266-179858840 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C5orf45 | NM_001017987.2 | NP_001017987.1 | ||
| NM_016175.3 | NP_057259.2 | |||
| AF153685.1 | AAD51611.1 | |||
| AK293901.1 | ||||
| AK295104.1 | 4 | 556 | ||
| AK298954.1 | ||||
| BC002739.2 | AAH02739.2 | |||
| BC069051.1 | AAH69051.1 | |||
| BF965313.1 | ||||
| BQ642639.1 | ||||
| BX537968.1 | CAD97932.1 | |||
| BX647375.1 | 6 | 696 | ||
| CR936785.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469579 | Chr.5:179757830 - 179933191 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map