Assay Details
Target Gene Details
Entrez Gene ID: | 4883 |
Gene Name: | natriuretic peptide receptor 3 |
Gene Aliases: |
ANP-C, ANPR-C, ANPRC, C5orf23, GUCY2B, NPR-C, NPRC |
Location: |
Chr.5:32689076-32791724 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPR3 | NM_001204376.1 | NP_001191305.1 | ||
| XM_005248309.1 | XP_005248366.1 | |||
| XM_011514047.1 | XP_011512349.1 | |||
| XM_011514048.2 | XP_011512350.1 | |||
| XM_011514049.2 | XP_011512351.1 | |||
| XM_011514050.2 | XP_011512352.1 | |||
| XM_017009492.1 | XP_016864981.1 | |||
| AK300094.1 | ||||
| AK308301.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1017141 | Chr.5:32615443 - 32722393 on Build GRCh38 | Gain |
|
| nsv950666 | Chr.5:32710695 - 32717394 on Build GRCh38 | Deletion |
|
| esv2759338 | Chr.5:32245255 - 32738538 on Build GRCh38 | Loss |
|
| esv3604616 | Chr.5:32625459 - 32725468 on Build GRCh38 | Gain |
|
| nsv329020 | Chr.5:32710875 - 32715346 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map