Assay Details
Target Gene Details
Entrez Gene ID: | 84250 |
Gene Name: | SMC5-SMC6 complex localization factor 1 |
Gene Aliases: |
ANKRD32, BRCTD1, BRCTx |
Location: |
Chr.5:94613477-94697733 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLF1 | NM_032290.3 | 4 | 416 | NP_115666.2 |
| XM_017009978.1 | 4 | 597 | XP_016865467.1 | |
| XM_017009979.1 | 4 | 5178 | XP_016865468.1 | |
| XM_017009980.1 | 4 | 598 | XP_016865469.1 | |
| XM_017009981.1 | 4 | 448 | XP_016865470.1 | |
| XM_017009982.1 | 4 | 597 | XP_016865471.1 | |
| AK297452.1 | 4 | 424 | ||
| AK299080.1 | 4 | 442 | ||
| AL050298.1 | 4 | 421 | CAB43397.2 | |
| BC054885.1 | 4 | 418 | AAH54885.2 | |
| BC070332.1 | 4 | 454 | AAH70332.1 | |
| BC107571.1 | 4 | 416 | AAI07572.1 | |
| CR936768.1 | 4 | 422 | ||
| DR001089.1 | 4 | 416 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv462254 | Chr.5:94621232 - 94721120 on Build GRCh38 | Loss |
|
| esv2759354 | Chr.5:94429500 - 94706496 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map