Assay Details
Target Gene Details
Entrez Gene ID: | 253314 |
Gene Name: | eukaryotic translation initiation factor 4E family member 1B |
Gene Aliases: |
- |
Location: |
Chr.5:176630613-176646644 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF4E1B | NM_001099408.1 | NP_001092878.1 | ||
| XM_011534499.2 | XP_011532801.1 | |||
| AK094270.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv600346 | Chr.5:176597879 - 176669235 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map