Assay Details
Target Gene Details
Entrez Gene ID: | 9509 |
Gene Name: | ADAM metallopeptidase with thrombospondin type 1 motif 2 |
Gene Aliases: |
ADAM-TS2, ADAMTS-2, ADAMTS-3, NPI, PC I-NP, PCI-NP, PCINP, PCPNI, PNPI |
Location: |
Chr.5:179110851-179345430 on Build GRCh38 |
Assay Gene Location: | Within Intron 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAMTS2 | NM_014244.4 | NP_055059.2 | ||
| XM_017010063.1 | XP_016865552.1 | |||
| AI417257.1 | ||||
| AJ003125.1 | CAA05880.1 | |||
| BX479109.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3607736 | Chr.5:179113082 - 179119734 on Build GRCh38 | Gain |
|
| nsv1033597 | Chr.5:178996100 - 179325094 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map