Assay Details
Target Gene Details
Entrez Gene ID: | 642366 |
Gene Name: | uncharacterized LOC642366 |
Gene Aliases: |
- |
Location: |
Chr.5:51372736-51383332 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC642366 | NR_046243.1 | 2 | 1474 | |
| CR936786.1 | 1 | 1537 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830288 | Chr.5:51302388 - 51459407 on Build GRCh38 | Gain |
|

Genomic Map