Assay Details
Target Gene Details
Entrez Gene ID: | 135152 |
Gene Name: | beta-1,3-glucuronyltransferase 2 |
Gene Aliases: |
GLCATS |
Location: |
Chr.6:70861366-70957085 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B3GAT2 | NM_080742.2 | NP_542780.1 | ||
| AB075843.1 | ||||
| AK055248.1 | BAB70889.1 | |||
| AK290018.1 | ||||
| AY070019.1 | AAL57718.1 | |||
| BC113995.1 | AAI13996.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525542 | Chr.6:70879806 - 70890184 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map