Assay Details
Target Gene Details
Entrez Gene ID: | 4835 |
Gene Name: | NAD(P)H quinone dehydrogenase 2 |
Gene Aliases: |
DHQV, DIA6, NMOR2, QR2 |
Location: |
Chr.6:2999816-3019876 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NQO2 | NM_000904.4 | NP_000895.2 | ||
| NM_001290221.1 | NP_001277150.1 | |||
| NM_001290222.1 | NP_001277151.1 | |||
| NM_001318940.1 | NP_001305869.1 | |||
| AK056981.1 | ||||
| AK311746.1 | ||||
| BI333292.1 | ||||
| BI670476.1 | ||||
| CD014013.1 | ||||
| J02888.1 | AAA60239.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv600833 | Chr.6:2970334 - 3010915 on Build GRCh38 | Loss |
|
| nsv1029300 | Chr.6:2997310 - 3024828 on Build GRCh38 | Loss |
|
| nsv5177 | Chr.6:2998306 - 3031324 on Build GRCh38 | Insertion |
|
| nsv830575 | Chr.6:2847376 - 3035682 on Build GRCh38 | Loss |
|
| esv2759404 | Chr.6:2989348 - 3015823 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map