Assay Details
Target Gene Details
Entrez Gene ID: | 112609 |
Gene Name: | melanocortin 2 receptor accessory protein 2 |
Gene Aliases: |
BMIQ18, C6orf117, bA51G5.2 |
Location: |
Chr.6:84032621-84146278 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRAP2 | NM_138409.2 | NP_612418.2 | ||
| XM_011535400.2 | XP_011533702.1 | |||
| XM_017010219.1 | XP_016865708.1 | |||
| XM_017010220.1 | XP_016865709.1 | |||
| XM_017010221.1 | XP_016865710.1 | |||
| AK090775.1 | BAC03517.1 | |||
| AK292736.1 | ||||
| AK307078.1 | ||||
| BC010003.2 | AAH10003.2 | |||
| BC039855.1 | AAH39855.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830719 | Chr.6:83930218 - 84117854 on Build GRCh38 | Gain |
|
| esv2759451 | Chr.6:84023580 - 84086850 on Build GRCh38 | Loss |
|
| nsv1025798 | Chr.6:83654421 - 84334553 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map