Assay Details
Target Gene Details
Entrez Gene ID: | 6239 |
Gene Name: | ras responsive element binding protein 1 |
Gene Aliases: |
FINB, HNT, LZ321, RREB-1, Zep-1 |
Location: |
Chr.6:7107597-7251980 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RREB1 | NM_001168344.1 | NP_001161816.1 | ||
| D49835.1 | BAA23165.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3890787 | Chr.6:6996928 - 7981438 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map