Assay Details
Target Gene Details
Entrez Gene ID: | 449520 |
Gene Name: | gametogenetin binding protein 1 (pseudogene) |
Gene Aliases: |
- |
Location: |
Chr.6:33583699-33589026 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GGNBP1 | NR_028361.1 | |||
| AY383626.1 | AAR26430.1 | |||
| AY383627.1 | AAR26431.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3608630 | Chr.6:33439052 - 34076935 on Build GRCh38 | Gain |
|
| dgv85n68 | Chr.6:33583425 - 33743569 on Build GRCh38 | Loss |
|
| nsv602833 | Chr.6:33567103 - 33620370 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map