Assay Details
Target Gene Details
Entrez Gene ID: | 401253 |
Gene Name: | long intergenic non-protein coding RNA 336 |
Gene Aliases: |
C6orf227, NCRNA00336 |
Location: |
Chr.6:33586106-33593338 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00336 | NR_027908.1 | |||
| AK125740.1 | BAC86269.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3608630 | Chr.6:33439052 - 34076935 on Build GRCh38 | Gain |
|
| dgv85n68 | Chr.6:33583425 - 33743569 on Build GRCh38 | Loss |
|
| nsv602833 | Chr.6:33567103 - 33620370 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs111357030] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map