Assay Details
Target Gene Details
Entrez Gene ID: | 154075 |
Gene Name: | sterile alpha motif domain containing 3 |
Gene Aliases: |
- |
Location: |
Chr.6:130133203-130365425 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SAMD3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv604663 | Chr.6:129482340 - 130671010 on Build GRCh38 | Gain |
|
| esv3610781 | Chr.6:130108614 - 130242086 on Build GRCh38 | Gain |
|

Genomic Map