Assay Details
Target Gene Details
Entrez Gene ID: | 389362 |
Gene Name: | proteasome assembly chaperone 4 |
Gene Aliases: |
C6orf86, PAC4, bA506K6.2 |
Location: |
Chr.6:3254325-3308224 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMG4 | NM_001128591.1 | NP_001122063.1 | ||
| NM_001128592.1 | NP_001122064.1 | |||
| NM_001135750.1 | 2 | 322 | NP_001129222.1 | |
| XM_011514591.2 | XP_011512893.1 | |||
| XM_011514593.2 | XP_011512895.1 | |||
| XM_011514594.2 | XP_011512896.1 | |||
| XM_011514595.2 | XP_011512897.1 | |||
| AA948231.1 | ||||
| AK095683.1 | ||||
| AK125314.1 | ||||
| AK126623.1 | 2 | 277 | ||
| BM738331.1 | ||||
| BX648600.1 | 1 | 214 | ||
| CN429483.1 | ||||
| DA093618.1 | 2 | 277 |
More Information
Set Membership: |
|

Genomic Map