Assay Details
Target Gene Details
Entrez Gene ID: | 100271873 |
Gene Name: | antisense of IGF2R non-protein coding RNA |
Gene Aliases: |
AIR, IGF2R-AS, IGF2R-AS1, IGF2RAS, NCRNA00088 |
Location: |
Chr.6:160003291-160007664 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AIRN | NR_047511.1 | |||
| NR_047514.1 | 1 | 2077 | ||
| AW008515.1 | 1 | 90 |
Target Gene Details
Entrez Gene ID: | 3482 |
Gene Name: | insulin like growth factor 2 receptor |
Gene Aliases: |
CD222, CI-M6PR, CIMPR, M6P-R, M6P/IGF2R, MPR 300, MPR1, MPR300, MPRI |
Location: |
Chr.6:159969099-160111504 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGF2R | NM_000876.3 | NP_000867.2 | ||
| AB209668.1 | BAD92905.1 | |||
| J03528.1 | AAA59866.1 | |||
| Y00285.1 | CAA68395.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv35098 | Chr.6:160001557 - 160649838 on Build GRCh38 | Gain |
|
| nsv1074441 | Chr.6:160005567 - 160006268 on Build GRCh38 | Deletion |
|
| esv2759484 | Chr.6:159951212 - 160813633 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map