Assay Details
Target Gene Details
Entrez Gene ID: | 100130890 |
Gene Name: | thiosulfate sulfurtransferase (rhodanese)-like domain containing 3 |
Gene Aliases: |
- |
Location: |
Chr.6:99424854-99579288 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSTD3 | XM_017010141.1 | XP_016865630.1 |
Target Gene Details
Entrez Gene ID: | 85015 |
Gene Name: | ubiquitin specific peptidase 45 |
Gene Aliases: |
- |
Location: |
Chr.6:99432306-99515770 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP45 | NM_001080481.1 | 17 | 2769 | NP_001073950.1 |
| XM_005267169.3 | 17 | 2372 | XP_005267226.1 | |
| XM_005267170.4 | 17 | 2341 | XP_005267227.1 | |
| XM_005267172.3 | XP_005267229.1 | |||
| XM_011536200.2 | 17 | 3599 | XP_011534502.1 | |
| XM_017011381.1 | 8 | 1652 | XP_016866870.1 | |
| XM_017011382.1 | 16 | 2674 | XP_016866871.1 | |
| XM_017011383.1 | 16 | 2200 | XP_016866872.1 | |
| XM_017011384.1 | 17 | 2314 | XP_016866873.1 | |
| AJ583819.2 | 16 | 2236 | CAE47746.2 | |
| AL713747.2 | 12 | 1830 | CAD89915.1 | |
| AL832030.1 | 10 | 1495 | CAD91148.1 | |
| AL832037.1 | 4 | 294 | CAI46150.1 | |
| AL832040.1 | 4 | 294 | CAI46152.1 | |
| BC150648.2 | 17 | 2336 | ||
| BC157838.1 | 17 | 2336 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv604254 | Chr.6:99434687 - 99473946 on Build GRCh38 | Loss |
|
| nsv1029536 | Chr.6:98422726 - 99818247 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map