Assay Details
Target Gene Details
Entrez Gene ID: | 94026 |
Gene Name: | POM121 transmembrane nucleoporin like 2 |
Gene Aliases: |
POM121-L, POM121L |
Location: |
Chr.6:27309063-27312232 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POM121L2 | NM_033482.3 | 1 | 847 | NP_258443.2 |
More Information
Set Membership: |
|

Genomic Map