Assay Details
Target Gene Details
Entrez Gene ID: | 9749 |
Gene Name: | phosphatase and actin regulator 2 |
Gene Aliases: |
C6orf56 |
Location: |
Chr.6:143608180-143831185 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHACTR2 | NM_001100166.1 | NP_001093636.1 | ||
| NM_014721.2 | NP_055536.2 | |||
| AB014580.1 | ||||
| AK300221.1 | ||||
| BC150630.1 | ||||
| BC152416.1 | ||||
| DC381966.1 | ||||
| DN993504.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5512 | Chr.6:143603100 - 143634776 on Build GRCh38 | Insertion |
|
| nsv1032369 | Chr.6:143604237 - 143686376 on Build GRCh38 | Gain |
|
| esv2759476 | Chr.6:143595484 - 143758839 on Build GRCh38 | Loss |
|
| nsv604805 | Chr.6:143609267 - 143684812 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map