Assay Details
Target Gene Details
Entrez Gene ID: | 5425 |
Gene Name: | DAN polymerase delta 2, accessory subunit |
Gene Aliases: |
- |
Location: |
Chr.7:44114680-44123668 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLD2 | NM_001127218.2 | NP_001120690.1 | ||
| NM_001256879.1 | 1 | 210 | NP_001243808.1 | |
| NM_006230.3 | NP_006221.2 | |||
| XM_011515431.1 | XP_011513733.1 | |||
| AK292347.1 | 1 | 210 | ||
| AK298168.1 | ||||
| AK312294.1 | ||||
| AU117477.2 | ||||
| BC000459.2 | AAH00459.1 | |||
| BP224472.1 | ||||
| DB025069.1 | 1 | 172 | ||
| U21090.1 | AAC50216.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830983 | Chr.7:44068842 - 44201624 on Build GRCh38 | Loss |
|
| esv998294 | Chr.7:44122163 - 44132378 on Build GRCh38 | Deletion |
|
| nsv8082 | Chr.7:43956156 - 44207229 on Build GRCh38 | Gain |
|
| nsv428165 | Chr.7:43855828 - 44172960 on Build GRCh38 | Gain |
|
| nsv470210 | Chr.7:44045936 - 44139229 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map