Assay Details
Target Gene Details
Entrez Gene ID: | 55607 |
Gene Name: | protein phosphatase 1 regulatory subunit 9A |
Gene Aliases: |
NRB1, NRBI, Neurabin-I |
Location: |
Chr.7:94907202-95296415 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422248 | Chr.7:94543833 - 94955318 on Build GRCh38 | Duplication |
|
| nsv1023193 | Chr.7:93602376 - 95828660 on Build GRCh38 | Gain |
|
| dgv11513n54 | Chr.7:94709551 - 94942020 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map