Assay Details
Target Gene Details
Entrez Gene ID: | 10344 |
Gene Name: | C-C motif chemokine ligand 26 |
Gene Aliases: |
IMAC, MIP-4a, MIP-4alpha, SCYA26, TSC-1 |
Location: |
Chr.7:75769524-75791583 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCL26 | NM_006072.4 | NP_006063.1 | ||
| XM_017011672.1 | XP_016867161.1 | |||
| AY358893.1 | AAQ89252.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain |
|
| nsv518028 | Chr.7:75539242 - 75825134 on Build GRCh38 | Gain |
|
| esv3613739 | Chr.7:75787266 - 75791244 on Build GRCh38 | Loss |
|
| dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion |
|
| esv2764051 | Chr.7:75663839 - 75875518 on Build GRCh38 | Gain+Loss |
|
| esv3613726 | Chr.7:75699055 - 75796413 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map