Assay Details
Target Gene Details
Entrez Gene ID: | 114796 |
Gene Name: | PSMG3 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.7:1570073-1589626 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMG3-AS1 | NR_021487.2 | |||
| NR_027328.1 | ||||
| NR_027329.1 | ||||
| AB067495.1 | BAB67801.1 | |||
| AK054665.1 | ||||
| BX538028.1 | ||||
| DA046883.1 | ||||
| DA928000.1 | ||||
| DB075169.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950786 | Chr.7:1381265 - 1675664 on Build GRCh38 | Deletion |
|
| esv2759499 | Chr.7:1159459 - 1650311 on Build GRCh38 | Loss |
|
| nsv830879 | Chr.7:1493586 - 1650311 on Build GRCh38 | Gain+Loss |
|
| nsv5615 | Chr.7:1528557 - 1573208 on Build GRCh38 | Deletion |
|
| nsv605820 | Chr.7:1518331 - 1574625 on Build GRCh38 | Loss |
|

Genomic Map