Assay Details
Target Gene Details
Entrez Gene ID: | 155465 |
Gene Name: | anterior gradient 3, protein disulphide isomerase family member |
Gene Aliases: |
AG-3, AG3, BCMP11, HAG3, PDIA18, hAG-3 |
Location: |
Chr.7:16854711-16881989 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGR3 | NM_176813.4 | NP_789783.1 | ||
| XM_005249627.2 | XP_005249684.1 | |||
| XM_011515152.2 | XP_011513454.1 | |||
| AY069977.1 | AAL55402.1 | |||
| AY359065.1 | AAQ89424.1 | |||
| BC058284.1 | AAH58284.1 | |||
| BM984550.1 | ||||
| BU682778.1 | ||||
| CB242846.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3571672 | Chr.7:16860947 - 16875799 on Build GRCh38 | Loss |
|
| esv3891076 | Chr.7:16794737 - 17707004 on Build GRCh38 | Gain |
|
| esv2752152 | Chr.7:16824103 - 16906601 on Build GRCh38 | Loss |
|
| nsv1027284 | Chr.7:16810362 - 16968543 on Build GRCh38 | Loss |
|
| dgv6289n100 | Chr.7:16230707 - 17452994 on Build GRCh38 | Gain |
|
| nsv606339 | Chr.7:16813184 - 16964677 on Build GRCh38 | Loss |
|
| esv3612405 | Chr.7:16859996 - 16877613 on Build GRCh38 | Loss |
|
| esv2761305 | Chr.7:16859573 - 16875812 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map