Assay Details
Target Gene Details
Entrez Gene ID: | 28988 |
Gene Name: | drebrin like |
Gene Aliases: |
ABP1, HIP-55, HIP55, SH3P7 |
Location: |
Chr.7:44044640-44061716 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DBNL | NM_001014436.2 | NP_001014436.1 | ||
| NM_001122956.1 | NP_001116428.1 | |||
| NM_001284313.1 | NP_001271242.1 | |||
| NM_001284315.1 | NP_001271244.1 | |||
| NM_014063.6 | NP_054782.2 | |||
| XM_011515325.1 | XP_011513627.1 | |||
| AB209486.1 | BAD92723.1 | |||
| AF077353.1 | AAF80228.1 | |||
| AF151364.1 | AAG13120.1 | |||
| AF197060.1 | AAF13701.1 | |||
| AF218020.3 | AAG17262.2 | |||
| AF250287.1 | AAF81273.1 | |||
| AK027367.1 | BAB55065.1 | |||
| AK293151.1 | ||||
| AK293279.1 | ||||
| AK293340.1 | ||||
| AK293698.1 | ||||
| AK296778.1 | ||||
| AK298102.1 | ||||
| AK300630.1 | ||||
| AK302037.1 | ||||
| BC011677.2 | AAH11677.1 | |||
| BC031687.1 | AAH31687.1 | |||
| CR457167.1 | CAG33448.1 | |||
| DC404852.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv8082 | Chr.7:43956156 - 44207229 on Build GRCh38 | Gain |
|
| nsv428165 | Chr.7:43855828 - 44172960 on Build GRCh38 | Gain |
|
| nsv470210 | Chr.7:44045936 - 44139229 on Build GRCh38 | Loss |
|
| nsv830980 | Chr.7:43865998 - 44078539 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map