Assay Details
Target Gene Details
Entrez Gene ID: | 5447 |
Gene Name: | cytochrome p450 oxidoreductase |
Gene Aliases: |
CPR, CYPOR, P450R |
Location: |
Chr.7:75915102-75986855 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POR | NM_000941.2 | NP_000932.3 | ||
| AK129978.1 | ||||
| AK290529.1 | ||||
| AK312543.1 | ||||
| BP244929.1 | ||||
| BX648619.1 | CAH56151.1 | |||
| DA701694.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain |
|
| esv3613742 | Chr.7:75912784 - 76036530 on Build GRCh38 | Loss |
|
| dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map