Assay Details
Target Gene Details
Entrez Gene ID: | 4698 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit A5 |
Gene Aliases: |
B13, CI-13KD-B, CI-13kB, NUFM, UQOR13 |
Location: |
Chr.7:123536997-123601651 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFA5 | NM_001282419.2 | 4 | 2738 | NP_001269348.1 |
| NM_001282420.2 | 5 | 2800 | NP_001269349.1 | |
| NM_001282421.2 | 5 | 2791 | NP_001269350.1 | |
| NM_001282422.2 | 4 | 2666 | NP_001269351.1 | |
| NM_001291304.1 | 7 | 2977 | NP_001278233.1 | |
| NM_005000.4 | 5 | 2783 | NP_004991.1 | |
| NR_104168.2 | 6 | 2992 | ||
| NR_104169.2 | 6 | 2943 | ||
| NR_111925.1 | 5 | 2781 | ||
| NR_111926.1 | 8 | 2941 | ||
| CR933664.1 | 5 | 2800 | CAI45962.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524192 | Chr.7:122763367 - 123578468 on Build GRCh38 | Gain |
|
| nsv831116 | Chr.7:123447558 - 123619922 on Build GRCh38 | Gain+Loss |
|
| esv3891198 | Chr.7:121885430 - 123886905 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map