Assay Details
Target Gene Details
Entrez Gene ID: | 64921 |
Gene Name: | CAS1 domain containing 1 |
Gene Aliases: |
C7orf12, NBLA04196 |
Location: |
Chr.7:94509858-94633465 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CASD1 | XM_011516495.2 | XP_011514797.1 |
Target Gene Details
Entrez Gene ID: | 8910 |
Gene Name: | sarcoglycan epsilon |
Gene Aliases: |
DYT11, ESG |
Location: |
Chr.7:94585224-94656209 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422248 | Chr.7:94543833 - 94955318 on Build GRCh38 | Duplication |
|
| nsv1023193 | Chr.7:93602376 - 95828660 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map