Assay Details
Target Gene Details
Entrez Gene ID: | 222183 |
Gene Name: | serine/arginine repetitive matrix 3 |
Gene Aliases: |
- |
Location: |
Chr.7:76201893-76287292 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRRM3 | NM_001110199.2 | NP_001103669.1 | ||
| NM_001291831.1 | NP_001278760.1 | |||
| AI815474.1 | ||||
| CA949697.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain |
|
| dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs143855473] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map