Assay Details
Target Gene Details
Entrez Gene ID: | 3757 |
Gene Name: | potassium voltage-gated channel subfamily H member 2 |
Gene Aliases: |
ERG-1, ERG1, H-ERG, HERG, HERG1, Kv11.1, LQT2, SQT1 |
Location: |
Chr.7:150944956-150978314 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNH2 | NM_000238.3 | NP_000229.1 | ||
| NM_001204798.1 | NP_001191727.1 | |||
| NM_172056.2 | NP_742053.1 | |||
| NM_172057.2 | NP_742054.1 | |||
| XM_011516185.2 | XP_011514487.1 | |||
| XM_011516186.2 | XP_011514488.1 | |||
| XM_017012195.1 | XP_016867684.1 | |||
| XM_017012196.1 | XP_016867685.1 | |||
| AB044806.1 | BAB19682.1 | |||
| AF363636.1 | AAL37559.1 | |||
| AJ512214.1 | CAD54447.1 | |||
| AJ609614.1 | CAE82156.1 | |||
| BC001914.1 | AAH01914.2 | |||
| BC127673.1 | ||||
| BM674996.1 | ||||
| BT007336.1 | AAP36000.1 | |||
| DQ525913.1 | ||||
| FJ938021.1 | ||||
| U04270.1 | AAA62473.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1030109 | Chr.7:150615828 - 151003922 on Build GRCh38 | Gain |
|
| nsv465213 | Chr.7:150909200 - 150970122 on Build GRCh38 | Loss |
|
| dgv11721n54 | Chr.7:150901404 - 150970122 on Build GRCh38 | Loss |
|
| nsv951648 | Chr.7:150931413 - 151106413 on Build GRCh38 | Deletion |
|
| nsv608990 | Chr.7:150264185 - 151010400 on Build GRCh38 | Gain |
|
| esv2759574 | Chr.7:150841733 - 150966609 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs35117381] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map