Assay Details
Target Gene Details
Entrez Gene ID: | 100216545 |
Gene Name: | KMT2E antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.7:105010542-105014141 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KMT2E-AS1 | NR_024586.1 | 1 | 1111 | |
| AK094372.1 | 1 | 598 | ||
| AK123476.1 | 1 | 1053 | ||
| AK124046.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv608068 | Chr.7:104981955 - 105117036 on Build GRCh38 | Loss |
|
| nsv436902 | Chr.7:104802439 - 105205347 on Build GRCh38 | Insertion |
|

Genomic Map