Assay Details
Target Gene Details
Entrez Gene ID: | 93432 |
Gene Name: | maltase-glucoamylase 2 (putative) |
Gene Aliases: |
- |
Location: |
Chr.7:142111703-142223733 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 14 - Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MGAM2 | NM_001293626.1 | NP_001280555.1 | ||
| XM_011516692.2 | XP_011514994.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv608590 | Chr.7:142136069 - 142240255 on Build GRCh38 | Gain |
|
| nsv1035111 | Chr.7:142136794 - 142310364 on Build GRCh38 | Gain |
|
| nsv465131 | Chr.7:142072828 - 142629318 on Build GRCh38 | Loss |
|
| esv34021 | Chr.7:142139692 - 142182591 on Build GRCh38 | Loss |
|
| esv2759568 | Chr.7:142010611 - 142229156 on Build GRCh38 | Gain+Loss |
|
| esv33962 | Chr.7:142061773 - 142348667 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map