Assay Details
Target Gene Details
Entrez Gene ID: | 64921 |
Gene Name: | CAS1 domain containing 1 |
Gene Aliases: |
C7orf12, NBLA04196 |
Location: |
Chr.7:94509858-94633465 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CASD1 | XM_011516495.2 | XP_011514797.1 |
Target Gene Details
Entrez Gene ID: | 8910 |
Gene Name: | sarcoglycan epsilon |
Gene Aliases: |
DYT11, ESG |
Location: |
Chr.7:94585224-94656209 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SGCE | NM_001099400.1 | NP_001092870.1 | ||
| NM_001099401.1 | NP_001092871.1 | |||
| NM_001301139.1 | NP_001288068.1 | |||
| NM_003919.2 | NP_003910.1 | |||
| XM_005250675.4 | XP_005250732.1 | |||
| XM_005250677.4 | XP_005250734.1 | |||
| XM_011516663.2 | 12 | 1860 | XP_011514965.1 | |
| XM_011516664.2 | 11 | 1833 | XP_011514966.1 | |
| XM_011516665.2 | 11 | 1755 | XP_011514967.1 | |
| XM_011516666.2 | 10 | 1728 | XP_011514968.1 | |
| XM_011516667.2 | 13 | 1892 | XP_011514969.1 | |
| XM_011516669.2 | 12 | 1798 | XP_011514971.1 | |
| XM_017012763.1 | 12 | 1728 | XP_016868252.1 | |
| XM_017012764.1 | XP_016868253.1 | |||
| XM_017012765.1 | XP_016868254.1 | |||
| XM_017012766.1 | XP_016868255.1 | |||
| XM_017012767.1 | 11 | 1784 | XP_016868256.1 | |
| XM_017012768.1 | XP_016868257.1 | |||
| XM_017012769.1 | XP_016868258.1 | |||
| AB117974.1 | BAD21206.1 | |||
| AF031920.1 | AAC14021.1 | |||
| AF036364.1 | AAC04368.1 | |||
| AF516515.1 | AAM64204.1 | |||
| AJ000534.1 | CAA04167.1 | |||
| AK298220.1 | ||||
| AK313438.1 | ||||
| AL547747.3 | ||||
| AY359042.1 | AAQ89401.1 | |||
| BC021709.2 | AAH21709.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422248 | Chr.7:94543833 - 94955318 on Build GRCh38 | Duplication |
|
| nsv1023193 | Chr.7:93602376 - 95828660 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map